Amplite® Fluorimetric Sodium Ion Quantification Kit
Lieferant: AAT BIOQUEST
21325
AATB21325EA 687 EUR
AATB21325
Amplite® Fluorimetric Sodium Ion Quantification Kit
Tests Zelltests
Designed for consistent and dependable results. Amplite® fluorimetric sodium ion quantification kit uses our robust sodium ion indicator dye, SoNa™ 520, which exhibit great fluorescence intensity enhancement upon binding to sodium ions.
  • Robust protocol
  • SoNa™ 520 is perhaps the most robust sodium ion indicator with high selectivity
  • 12 month shelf life
This microplate-based assay kit requires extremely small amount of sample, it is particularly suitable for the determination of sodium ion concentration in microvolume format.
Lieferumfang: Shipped at RT.
Bitte beachten Research use only.