Portelite™ Fluorimetric Sodium Ion Quantification Kit
Lieferant: AAT BIOQUEST
Designed for consistent and dependable results. Portelite™ fluorimetric sodium ion quantification kit uses our robust sodium ion indicator dye, SoNa™ 520, which exhibit great fluorescence intensity enhancement upon binding to sodium ions.
- Robust protocol
- This nanodrop-based assay kit requires a small amount of sample, it is particularly suitable for the determination of sodium ion concentration in fields or on site
- 12-month shelf life
SoNa™ 520 is perhaps the best sodium ion indicator for rapidly determining sodium ion concentration in combination with a fluorescence device such as a fluorescence Nanodrop spectrophotometer or a fluorescence microplate reader.
Lieferumfang: Shipped at RT.
Bitte beachten Research use only.