8 CPG [TQ8 CPG], Tide Quencher™ 1000 Å
Lieferant: AAT BIOQUEST
2133
AATB2133EA
275
EUR
AATB2133
8 CPG [TQ8 CPG], Tide Quencher™ 1000 Å
8 CPG [TQ8 CPG]
Tide Quencher™ 8 (TQ8) is a non-fluorescent molecule designed to efficiently quench the fluorescence of IR fluorophores such as ICG, iFluor® 820, iFluor® 840 and iFluor® 860.
- Stable for 12 months
- Optimized for FRET applications
Quality tested in-house