Amplite® Fluorimetric Lithium Ion Quantification Kit
Lieferant: AAT BIOQUEST
21351
AATB21351EA
687
EUR
AATB21351
Amplite® Fluorimetric Lithium Ion Quantification Kit
Tests
Zelltests
Designed for consistent and dependable results. Amplite® fluorimetric lithium ion quantification kit uses our new robust lithium-ion indicator dye, Lithiumighty™ 520, which exhibits great fluorescence intensity enhancement upon binding to lithium ions.
- Lithiumighty ™ 520 is perhaps the most robust lithium-ion indicator with high selectivity
- Requires very small amount of sample
- 12 month shelf life
Microplate-based assay kit is particularly suitable for the determination of lithium ion concentration in a microvolume format.
Lieferumfang: Shipped at RT.
Bitte beachten Research use only.