8WS alkyne [TQ8WS alkyne], Tide Quencher™
Lieferant: AAT BIOQUEST
2137
AATB2137EA 309 EUR
AATB2137
8WS alkyne [TQ8WS alkyne], Tide Quencher™
8WS alkyne [TQ8WS alkyne]
Tide Quencher™ 8WS (TQ8WS) is a non-fluorescent molecule designed to efficiently quench the fluorescence of IR fluorophores such as ICG, iFluor® 820, iFluor® 840 and iFluor® 860.
  • Stable for 12 months
  • Optimized for FRET applications
Quality tested in-house
Bitte beachten For Research Use Only