RH 237 [N-(4-Sulphobutyl)-4-(6-(4-(dibutylamino)phenyl)hexatrienyl)pyridinium inner salt]
Lieferant: AAT BIOQUEST
21480
AATB21480EA 152 EUR
AATB21480
RH 237 [N-(4-Sulphobutyl)-4-(6-(4-(dibutylamino)phenyl)hexatrienyl)pyridinium inner salt]
RH 237 [N-(4-Sulphobutyl)-4-(6-(4-(dibutylamino)phenyl)hexatrienyl)pyridinium inner salt]
RH 237, also called N-(4-sulfobutyl)-4-(6-(4-(dibutylamino)phenyl)hexatrienyl)pyridinium, is a neuron-tracing dye.
  • Well-cited dye
  • Multiple applications
Bitte beachten For research use only.