RH421 ≥90%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
4-{4-[4-(Dipentylamino)phenyl]-1,3-butadienyl}-1-(4-sulphobutyl)pyridinium hydroxide, N-(4-sulphobutyl)-4-(4-(4-(dipentylamino)phenyl)butadienyl) pyridinium, inner salt
21482
AATB21482EA
152
EUR
AATB21482
RH421 ≥90%
RH421
RH 421, also called N-(4-Sulfobutyl)-4-(4-(4- (dipentylamino)phenyl)butadienyl)pyridinium, is a neuron-tracing dye.
Formel:
C₂₉H₄₂N₂O₃S Molecular Weight: 498,72 g/mol |
CAS-Nummer:
107610-19-5 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...