N-(3-Triethylammoniumpropyl)-4-(4-(dibutylamino)styryl)pyridinium dibromide (MM 1-43) ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
Pyridinium, 4-[2-[4-(dibutylamino)phenyl]ethenyl]- 1-[3-(triethylammonio)propyl]- , dibromide, SynaptoGreen™ C4, FM1-43
21483
AATB21483EA
152
EUR
AATB21483
N-(3-Triethylammoniumpropyl)-4-(4-(dibutylamino)styryl)pyridinium dibromide (MM 1-43) ≥95%
N-(3-Triethylammoniumpropyl)-4-(4-(dibutylamino)styryl)pyridinium dibromide (MM 1-43)
MM 1-43 is the abbreviation of our Membrane Marker 1-43, chemically called N-(3-triethylammoniumpropyl)-4-(4-(dibutylamino)styryl)pyridinium dibromide.
Formel:
C₃₀H₄₉Br₂N₃ Molecular Weight: 611,54 g/mol |
MDL:
MFCD00467815 CAS-Nummer: 149838-22-2 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...