RH 414 ≥90%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: N-(3-Triethylammoniumpropyl)-4-(4-(4-(diethylamino)phenyl)butadienyl)pyridinium dibromide

21485
AATB21485EA 230 EUR
AATB21485
RH 414 ≥90%
RH 414

RH 414 [N-(3-Triethylammoniumpropyl)-4-(4-(4-(diethylamino)phenyl)butadienyl)pyridinium dibromide] is a neuron-tracing dye.

Formel: C₂₈H₄₃Br₂N₃
Molecular Weight: 581,476 g/mol
CAS-Nummer: 161433-30-3
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR