N-(3-Triethylammoniumpropyl)-4-(6-(4-(diethylamino)phenyl)hexatrienyl)pyridinium dibromide (MM 4-64)

Lieferant: AAT BIOQUEST

Synonyme: FM® 4-64, Pyridinium, 4-[6-[4-(diethylamino)phenyl]- 1,3,5-hexatrienyl]-1-[3-(triethylammonio) propyl]-, dibromide, SynaptoRed™, MM 4-64

21487
AATB21487EA 307 EUR
AATB21487
N-(3-Triethylammoniumpropyl)-4-(6-(4-(diethylamino)phenyl)hexatrienyl)pyridinium dibromide (MM 4-64)
N-(3-Triethylammoniumpropyl)-4-(6-(4-(diethylamino)phenyl)hexatrienyl)pyridinium dibromide (MM 4-64)

MM 4-64 (FM 4-64) is the abbreviation of our Membrane Marker 4-64, chemically called N-(3-triethylammoniumpropyl)-4-(6-(4-(diethylamino)phenyl)hexatrienyl)pyridinium dibromide.


  • Lipophilic vital stain
  • Adaptable and robust
  • Multiple applications


In literature this membrane marker is also called 'FM 4-64' (FM® is the trademark of Molecular Probes). It is a lipophilic styryl dye that is used as a vital stain to follow bulk membrane-internalization and transport to the vacuole in yeast. MM 4-64 is a sensitive reporter of vacuolar dynamics, detecting such events as segregation structure formation during mitosis, vacuole fission/fusion events, and vacuolar morphology in different classes of vacuolar protein sorting mutants. It can be used for detecting endosome to vacuole membrane transport in vitro.


Bitte beachten For research use only.

Formel: C₃₀H₄₅Br₂N₃
Molecular Weight: 607,51 g/mol
MDL: MFCD00467819
CAS-Nummer: 162112-35-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR