MM 2-10 [N-(3-Triethylammoniumpropyl)-4-(4-(diethylamino)styryl)pyridinium dibromide]
Lieferant: AAT BIOQUEST
21489
AATB21489EA 152 EUR
AATB21489
MM 2-10 [N-(3-Triethylammoniumpropyl)-4-(4-(diethylamino)styryl)pyridinium dibromide]
MM 2-10 [N-(3-Triethylammoniumpropyl)-4-(4-(diethylamino)styryl)pyridinium dibromide]
MM 2-10 (FM 2-10) is the abbreviation of our Membrane Marker 2-10, chemically called N-(3-triethylammoniumpropyl)-4-(4-(diethylamino)styryl)pyridinium dibromide.
  • Lipophilic vital stain
  • Adaptable and Robust
  • Multiple applications
Bitte beachten For research use only.