N,N,N-Trimethyl-4-(6-phenyl-1,3,5-hexatrien-1-yl)phenylammonium-p-toluenesulphonate ≥95%

Lieferant: AAT BIOQUEST
Achtung

21492
AATB21492EA 152 EUR
AATB21492
N,N,N-Trimethyl-4-(6-phenyl-1,3,5-hexatrien-1-yl)phenylammonium-p-toluenesulphonate ≥95%
N,N,N-Trimethyl-4-(6-phenyl-1,3,5-hexatrien-1-yl)phenylammonium-p-toluenesulphonate

TMA-DPH is a fluorescent probe for membrane fluidity measurements.

Formel: C₂₈H₃₁NO₃S
Molecular Weight: 461,62 g/mol
CAS-Nummer: 115534-33-3
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR