Di-8-ANEPPS ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 4-(2-[6-(Dioctylamino)-2-naphthalenyl]ethenyl)-1-(3-sulfopropyl)pyridinium inner salt

21497
AATB21497EA 307 EUR
AATB21497
Di-8-ANEPPS ≥95%
Di-8-ANEPPS

Di-8-ANEPPS is widely used for monitoring fast membrane potential changes.

Formel: C₃₆H₅₂N₂O₃S
Molecular Weight: 592,87 g/mol
MDL: MFCD03453608
CAS-Nummer: 157134-53-7
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR