Di-4-ANEPPS ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 4-(2-(6-(Dibutylamino)-2-naphthalenyl)ethenyl)-1-(3-sulphopropyl)pyridinium hydroxide inner salt

21498
AATB21498EA 307 EUR
AATB21498
Di-4-ANEPPS ≥95%
Di-4-ANEPPS

Potential-sensitive ANEP dyes are zwitterionic molecule, exhibiting the most consistent potentiometric response in different cell and tissue types.

Formel: C₂₈H₃₆N₂O₃S
Molecular Weight: 480,66 g/mol
MDL: MFCD03453609
CAS-Nummer: 90134-00-2
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR