OxiVision™ Green Hydrogenperoxid-Sensor
Lieferant: AAT BIOQUEST
21505
AATB21505EA 152 EUR
AATB21505
OxiVision™ Green Hydrogenperoxid-Sensor
OxiVision™ Green
Despite the importance of H₂O₂ to human health and disease, the molecular mechanisms of its production, accumulation, trafficking, and function are insufficiently understood due to the lack of sensitive and specific H₂O₂ sensors that can be used in live cells.
  • Suitable for live cells
  • Extremely selective for Hydrogen peroxide
  • Bright signal
Bitte beachten For research use only.