Thiolite™ Blue, AM
Lieferant: AAT Bioquest
21506
AATB21506EA 152 EUR
AATB21506
Thiolite™ Blue, AM
Thiolite™ Blue, AM
Thiolite™ Blue is one of the most sensitive sensors for measuring thiol compounds.
  • No wash step required
  • Cell permeable
  • Improved sensitivity and signal
  • Optimized for intracellular detection
  • Multiple applications
Bitte beachten For research use only.