6-NED azide fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

217
AATB217EA 439 EUR
AATB217
6-NED azide fluoreszierende Probe
6-NED azide

5'-Labeled primer pairs include a fluorescently labeled oligo with a choice of dye on the 5' end, along with an unlabeled oligo, in each pair.


  • Alkyne-reactive probe

Molecular Weight: 615.78 g/mol
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1389aaba1c915bdc45c58a31f3a83a63ebfa677&vid=f7361e750b4995c7bd3d30735746926e70fb8471&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR