Native Dextran (from Eukaryotic cells), FITC (Fluorescein Isothiocyanate)
Lieferant: AAT BIOQUEST
21700
AATB21700EA
230
EUR
AATB21700
Native Dextran (from Eukaryotic cells), FITC (Fluorescein Isothiocyanate)
Proteine und Peptide
FITC-labeled dextran is used in cardiovascular, microcirculation, perfusion, cell monolayer and cell membrane permeability research as fluorescent flux tracer compound that supports the measurement of processes such as blood flow, membrane damage, vascular drainage and renal elimination.
- Well-retained in cells
- 20000 mw
- Multiple applications
Bitte beachten For research use only.