Dextran, M.W. 150,000 FITC labelled

Lieferant: AAT BIOQUEST

21704
AATB21704EA 218 EUR
AATB21704
Dextran, M.W. 150,000 FITC labelled
Dextran

FITC-labeled dextran is used in cardiovascular, microcirculation, perfusion, cell monolayer and cell membrane permeability research as fluorescent flux tracer compound that supports the measurement of processes such as blood flow, membrane damage, vascular drainage and renal elimination.


  • Well-retained in cells
  • 150000 mw
  • Multiple applications


From Eukaryotic cells.


Bitte beachten For research use only.

Formel: (C₆H₁₀O₅)n
MDL: MFCD00130935
CAS-Nummer: 9004-54-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR