TRITC-Dextran Conjugate (Average MW = ~70K)
Lieferant: AAT BIOQUEST
21708
AATB21708EA 292 EUR
AATB21708
TRITC-Dextran Conjugate (Average MW = ~70K)
Tests Zelltests
TRITC-labeled dextran is used in cardiovascular, microcirculation, perfusion, cell monolayer and cell membrane permeability research as fluorescent flux tracer compound that supports the measurement of processes such as blood flow, membrane damage, vascular drainage and renal elimination.
  • High quality
  • Multiple applications
Bitte beachten For research use only.