CytoTrace™ Ultra Green
Lieferant: AAT BIOQUEST
21800
AATB21800EA 383 EUR
AATB21800
CytoTrace™ Ultra Green
Tests Zelltests
CMFDA (5-chloromethylfluorescein diacetate) has been used as a prominent cell tracker in the research community for a long time.
  • Bright Signal
  • High Sensitivity
  • Fixable
  • Stable Signal
Bitte beachten For research use only.