CytoTrace™ Ultra Green
Lieferant: AAT BIOQUEST
21800
AATB21800EA
383
EUR
AATB21800
CytoTrace™ Ultra Green
Tests
Zelltests
CMFDA (5-chloromethylfluorescein diacetate) has been used as a prominent cell tracker in the research community for a long time.
- Bright Signal
- High Sensitivity
- Fixable
- Stable Signal
Bitte beachten For research use only.