Tide Quencher™ 1 azide, Tide Quencher™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TQ1 azide
2188
AATB2188EA
307
EUR
AATB2188
Tide Quencher™ 1 azide, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 1 azide
TQ1 is designed to be a superior quencher alternative to DABCYL.
- Stronger absorption
- Higher quenching efficiency
- Versatile reactive forms
- Excellent oligonucleotide and peptide labeling
Molecular Weight:
358,42 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...