Tide Quencher™ 1 alkyne, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ1 alkyne

2189
AATB2189EA 307 EUR
AATB2189
Tide Quencher™ 1 alkyne, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 1 alkyne

TQ1 is designed to be a superior quencher alternative to DABCYL.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 327,4 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7946346caa58286ee775638e37ae53bf6adb935e&vid=acdc90a9a8877e4674e411e2500c122feade5447&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR