Tide Quencher™ 1 acid, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ1 acid

2190
AATB2190EA 307 EUR
AATB2190
Tide Quencher™ 1 acid, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 1 acid

TQ1 is designed to be a superior quencher alternative to DABCYL.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 290,34 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46af941e13b5b91cf298f14189c2cd662cc38cc6&vid=15b71127fa277c0f4b91bd63d0989d4fb862e7d8&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR