Calcein Red™ sodium salt
Lieferant: AAT BIOQUEST
21901
AATB21901EA
307
EUR
AATB21901
Calcein Red™ sodium salt
Calcein Red™ sodium salt
Calcein AM is one of the most popular fluorescent probes used for labeling and monitoring cellular functions of live cells.
- In-house quality control
- long-wavelength emission signal
- Multiple applications
Bitte beachten For research use only.