Calcein Deep Red™
Lieferant: AAT BIOQUEST
21902
AATB21902EA
307
EUR
AATB21902
Calcein Deep Red™
Calcein Deep Red™
Calcein AM is one the most popular fluorescent probes used for labeling and monitoring cellular functions of live cells.
- In-house quality control
- Long-wavelength emission signal
- Multiple applications
Bitte beachten For research use only.