Calcein UltraBlue™ AM
Lieferant: AAT BIOQUEST
21908
AATB21908EA
307
EUR
AATB21908
Calcein UltraBlue™ AM
Calcein UltraBlue™ AM
Calcein UltraBlue™ AM is a cell-permeable version of Calcein UltraBlue™.
- Low cytotoxicity
- Excellent cell retention
- Cell permeable
- Suitable for multiplexing
Bitte beachten For research use only.