Calcein UltraBlue™ AM
Lieferant: AAT BIOQUEST
21908
AATB21908EA 307 EUR
AATB21908
Calcein UltraBlue™ AM
Calcein UltraBlue™ AM
Calcein UltraBlue™ AM is a cell-permeable version of Calcein UltraBlue™.
  • Low cytotoxicity
  • Excellent cell retention
  • Cell permeable
  • Suitable for multiplexing
Bitte beachten For research use only.