Calcein UltraBlue™ sodium salt
Lieferant: AAT BIOQUEST
21909
AATB21909EA
307
EUR
AATB21909
Calcein UltraBlue™ sodium salt
Calcein UltraBlue™ sodium salt
Calcein UltraBlue™ has similar spectral properties to those of Calcein Blue.
- Low cytotoxicity
- Improved photostability
- Brighter fluorescence
- Suitable for multiplexing
Bitte beachten For research use only.