Calcein UltraBlue™ sodium salt
Lieferant: AAT BIOQUEST
21909
AATB21909EA 307 EUR
AATB21909
Calcein UltraBlue™ sodium salt
Calcein UltraBlue™ sodium salt
Calcein UltraBlue™ has similar spectral properties to those of Calcein Blue.
  • Low cytotoxicity
  • Improved photostability
  • Brighter fluorescence
  • Suitable for multiplexing
Bitte beachten For research use only.