CypHer7E maleimide
Lieferant: AAT Bioquest
21915
AATB21915EA 673 EUR
AATB21915
CypHer7E maleimide
CypHer7E maleimide
CypHer7E maleimide is an excellent building block for developing site-specific pH probes.
  • Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only