Tide Quencher™ 1 CPG 500 Å, Tide Quencher™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TQ1 CPG
2193
AATB2193EA
273
EUR
AATB2193
Tide Quencher™ 1 CPG 500 Å, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 1 CPG
TQ1 is designed to be a superior quencher alternative to DABCYL.
- Stronger absorption
- Higher quenching efficiency
- Versatile reactive forms
- Excellent oligonucleotide and peptide labeling
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...