Tide Quencher™ 1 CPG 1000 Å, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ1 CPG

2194
AATB2194EA 273 EUR
AATB2194
Tide Quencher™ 1 CPG 1000 Å, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 1 CPG

TQ1 is designed to be a superior quencher alternative to DABCYL.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR