Tide Quencher™ 1 maleimide, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ1 maleimide

2196
AATB2196EA 307 EUR
AATB2196
Tide Quencher™ 1 maleimide, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 1 maleimide

TQ1 is designed to be a superior quencher alternative to DABCYL.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 412,46 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9dc4cc0420ed6985b6374de381829ac9afa502eb&vid=8f5f60f41eb24b9e8b985b944e59234def38837d&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR