Tide Quencher™ 1 phosphoramidite, Tide Quencher™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TQ1 phosphoramidite
2198
AATB2198EA
307
EUR
AATB2198
Tide Quencher™ 1 phosphoramidite, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 1 phosphoramidite
TQ1 is designed to be a superior quencher alternative to DABCYL.
- Stronger absorption
- Higher quenching efficiency
- Versatile reactive forms
- Excellent oligonucleotide and peptide labeling
Molecular Weight:
589,73 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...