Tide Quencher™ 1 succinimidyl ester, Tide Quencher™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TQ1 SE
2199
AATB2199EA
307
EUR
AATB2199
Tide Quencher™ 1 succinimidyl ester, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 1 succinimidyl ester
TQ1 is designed to be a superior quencher alternative to DABCYL.
- Stronger absorption
- Higher quenching efficiency
- Versatile reactive forms
- Excellent oligonucleotide and peptide labeling
Molecular Weight:
387,41 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...