6-VIC alkyne
Lieferant: AAT BIOQUEST
6-VIC alkyne is a convenient building block that is used to label a biomolecule that contains an azido group, such as azido-modified oligos.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only