Tide Quencher™ 2 acid, Tide Quencher™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TQ2 acid
2200
AATB2200EA
307
EUR
AATB2200
Tide Quencher™ 2 acid, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 2 acid
TQ2 is designed to be a superior quencher to FAM, HEX, TET, JOE, TF2 and rhodamine 6G.
- Stronger absorption
- Higher quenching efficiency
- Versatile reactive forms
- Excellent oligonucleotide and peptide labeling
Molecular Weight:
382,48 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...