Calcein tetraethyl acid ≥95%, Ultrareine Qualität

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 2,2',2'',2'''-(((3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-4',5'-diyl)bis(methylene))bis(azanetriyl))tetraacetic acid, Fluorescein-4',5'-bis(methyliminodiacetic acid), Calcein

22001
AATB22001EA 152 EUR
AATB22001
Calcein tetraethyl acid ≥95%, Ultrareine Qualität
Calcein tetraethyl acid

Calcein, also known as fluorexon, fluorescein complex, is a fluorescent calcium indicator with an excitation and emission wavelengths of 495 and 515 nm respectively.

Formel: C₃₀H₂₆N₂O₁₃
Molecular Weight: 622,53 g/mol
MDL: MFCD00075616
CAS-Nummer: 207124-64-9
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR