Calceinblau ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
4-Methylumbelliferonemethyleneiminodiacetic Acid, Umbelliferone Complexon
22006
AATB22006EA
120
EUR
AATB22006
Calceinblau ≥95%
Calcein blue
It has similar calcium binding properties to those of calcein.
Formel:
C₁₅H₁₅NO₇ Molecular Weight: 321,29 g/mol |
MDL:
MFCD00006867 CAS-Nummer: 54375-47-2 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...