Calcein blue AM ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: Calcein Blue Acetoxymethyl ester, N-[2-[(acetyloxy)methoxy]-2-oxoethyl]-N-[(7-hydroxy-4-methyl-2-oxo-2H-1-benzopyran-8-yl)methyl]-glycine, (acetyloxy)methyl ester

22007
AATB22007EA 120 EUR
AATB22007
Calcein blue AM ≥95%
Calcein blue AM

It is a cell-permeable version of calcein blue.

Formel: C₂₁H₂₃NO₁₁
Molecular Weight: 465,41 g/mol
CAS-Nummer: 168482-84-6
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR