Calcein Deep Red™ AM ester
Lieferant: AAT BIOQUEST
22011
AATB22011EA 307 EUR
AATB22011
Calcein Deep Red™ AM ester
Calcein Deep Red™ AM ester
Calcein AM is one the most popular fluorescent probes used for labeling and monitoring cellular functions of live cells.
  • Low cytotoxicity
  • Excellent cell retention
  • Cell permeable
  • Suitable for multiplexing
Bitte beachten For research use only.