CytoCalcein™ Violet 450 maximum excitation at 405 nm
Lieferant: AAT BIOQUEST
22012
AATB22012EA
307
EUR
AATB22012
CytoCalcein™ Violet 450 maximum excitation at 405 nm
CytoCalcein™ Violet 450
CytoCalcein™ Violet 450 is designed for labelling live cells in the same way to calcein, AM.
- Optimized for flow cytometry excitation
- Brighter signal
- Suitable for multiplexing
Bitte beachten For research use only.