CytoCalcein™ Violet 500 maximum excitation at 405 nm
Lieferant: AAT BIOQUEST
22013
AATB22013EA 307 EUR
AATB22013
CytoCalcein™ Violet 500 maximum excitation at 405 nm
CytoCalcein™ Violet 500
CytoCalcein™ Violet 500 is designed for labeling live cells in the same way to calcein, AM.
  • Optimized for flow cytometry excitation
  • Brighter signal
  • Suitable for Multiplexing
Bitte beachten For research use only.