Tide Quencher™ 2 amine, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ2 amine

2202
AATB2202EA 307 EUR
AATB2202
Tide Quencher™ 2 amine, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 2 amine

TQ2 is designed to be a superior quencher to FAM, HEX, TET, JOE, TF2 and rhodamine 6G.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 424,56 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f83d2e9f7e5af5b8b3776dfd1cd88f1a42732e2&vid=06a6374e3de28a7e8dd3c879eee7a32598c89c72&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR