Fluoresceindiacetat ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: Di-O-acetylfluorescein, 3,6-Diacetoxyfluoran, 3-Oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-diyl diacetate

22020
AATB22020EA 120 EUR
AATB22020
Fluoresceindiacetat ≥95%
Fluoresceindiacetat

FDA is a non-fluorescent hydrophobic fluorescein derivative that can pass through the cell membrane whereupon intracellular esterases hydrolyze the diacetate group producing the highly fluorescent product fluorescein.

Formel: C₂₄H₁₆O₇
Molecular Weight: 416,38 g/mol
MDL: MFCD00005062
CAS-Nummer: 596-09-8
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR