5(6)-Carboxyfluorescein diacetate Gemisch aus Isomeren ≥95%

Lieferant: AAT BIOQUEST

Synonyme: CFDA, 5(6)-Carboxy-di-O-acetylfluorescein, 5(6)-CFDA

22021
AATB22021EA 120 EUR
AATB22021
5(6)-Carboxyfluorescein diacetate Gemisch aus Isomeren ≥95%
5(6)-Carboxyfluorescein diacetate

It is widely recognized that fluorescent labeling of cells is an effective means to determine total cell numbers or how many viable cells exist in a sample.


  • Bright Fluorescence
  • Multiple Bioconjugation Applications


Bitte beachten For research use only.

Formel: C₂₅H₁₆O₉
Molecular Weight: 460.39 g/mol
MDL: MFCD00467494
CAS-Nummer: 124387-19-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR