5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)] ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: CFDA-SE, CFSE, Carboxyfluorescein diacetate succinimidyl ester, 5(6)-CFDA N-succinmidyl ester, 5(6)-Carboxyfluorescein diacetate succinimidyl ester, 5(6)-CFDA SE

22022
AATB22022EA 152 EUR
AATB22022
5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)] ≥95%
5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)]

It is widely recognized that fluorescent labeling of cells is an effective means to determine total cell numbers or how many viable cells exist in a sample.

Formel: C₂₉H₁₉NO₁₁
Molecular Weight: 557,467 g/mol
MDL: MFCD00037466
CAS-Nummer: 150347-59-4
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR