5(6)-Carboxy-2′,7′- dichlorofluorescein diacetate Gemisch aus Isomeren ≥95%
Lieferant: AAT BIOQUEST
Synonyme:
Carboxy-DCFDA, 5-(6)-Carboxydichlorofluorescein diacetate, 5(6)-CDCFDA, CDCFDA
22025
AATB22025EA
152
EUR
AATB22025
5(6)-Carboxy-2′,7′- dichlorofluorescein diacetate Gemisch aus Isomeren ≥95%
5(6)-Carboxy-2′,7′- dichlorofluorescein diacetate
It is widely recognized that fluorescent labeling of cells is an effective means to determine total cell numbers or how many viable cells exist in a sample.
- Bright Fluorescence
- Multiple Bioconjugation Applications
Bitte beachten For research use only.
Formel:
C₂₅H₁₄Cl₂O₉ Molecular Weight: 529,28 g/mol |
MDL:
MFCD08459585 CAS-Nummer: 127770-45-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...