5(6)-Carboxy-2′,7′-dichlorofluorescein diacetate N-succinimidyl ester (CDCFDA) Gemisch aus Isomeren ≥90%
Lieferant: AAT BIOQUEST
Synonyme:
Carboxy-2′,7′- dichlorofluorescein diacetate, succinimidyl ester, 5 and 6 isomers, Carboxy-DCFDA-N-succinimidyl ester, CDCFDA
22026
AATB22026EA
152
EUR
AATB22026
5(6)-Carboxy-2′,7′-dichlorofluorescein diacetate N-succinimidyl ester (CDCFDA) Gemisch aus Isomeren ≥90%
5(6)-Carboxy-2′,7′-dichlorofluorescein diacetate N-succinimidyl ester (CDCFDA)
It is widely recognized that fluorescent labeling of cells is an effective means to determine total cell numbers or how many viable cells exist in a sample.
- Bright Fluorescence
- Multiple Bioconjugation Applications
Bitte beachten For research use only.
Formel:
C₂₉H₁₇Cl₂NO₁₁ Molecular Weight: 626,35 g/mol |
CAS-Nummer:
147265-60-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...