4-(4-(Dihexadecylamino)styryl)-N-methylpyridinium iodide (DiA) ≥95%
Lieferant: AAT BIOQUEST
22030
AATB22030EA
120
EUR
AATB22030
4-(4-(Dihexadecylamino)styryl)-N-methylpyridinium iodide (DiA) ≥95%
4-(4-(Dihexadecylamino)styryl)-N-methylpyridinium iodide (DiA)
DiA is a lipophilic fluorescent stain for labeling membranes and other hydrophobic structures.
- Lipophilic
- Even Cellular Staining
Bitte beachten For research use only.
Formel:
C₄₆H₇₉N₂.I Molecular Weight: 787,04 g/mol |
CAS-Nummer:
114041-00-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...