1,1'-Dioctadecyl-3,3,3',3'-tetramethylindodicarbocyanine (DID) 5 mM in DMSO ≥95% für die Markierung

Lieferant: AAT BIOQUEST

22033
AATB22033EA 307 EUR
AATB22033
1,1'-Dioctadecyl-3,3,3',3'-tetramethylindodicarbocyanine (DID) 5 mM in DMSO ≥95% für die Markierung
1,1'-Dioctadecyl-3,3,3',3'-tetramethylindodicarbocyanine (DID)

DiI, DiO, DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures.


  • Lipophilic
  • Even Cellular Staining


Bitte beachten For research use only.

Formel: C₆₁H₉₉ClN₂O₄
Molecular Weight: 959,923 g/mol
CAS-Nummer: 127274-91-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR